ENA372572776 Enamine

5-cyano-3-methyl-1-benzothiophene-2-sulfonyl chloride

  • MDL number MFCD14706454
  • Cas Number : 1030857-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111400
Availability details for
Quantity
  • Availability
  • Add to Cart