ENA372572793 Enamine

4-(dimethylamino)-2-methoxybenzoic acid

  • MDL number MFCD06203180
  • Cas Number : 95420-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111428
Availability details for
Quantity
  • Availability
  • Add to Cart