ENA372572794 Enamine

1-(2-ethoxy-2-oxoethyl)-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD12157282
  • Cas Number : 1183616-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111429
Availability details for
Quantity
  • Availability
  • Add to Cart