ENA372572803 Enamine

1-(4,5,6,7-tetrahydro-1-benzothiophen-2-yl)ethan-1-one

  • MDL number MFCD01939673
  • Cas Number : 57021-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-36
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111440
Availability details for
Quantity
  • Availability
  • Add to Cart