ENA372572816 Enamine

5-methylpyrrolidin-3-ol

  • MDL number MFCD19665354
  • Cas Number : 94134-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111494
Availability details for
Quantity
  • Availability
  • Add to Cart