ENA372572827 Enamine

ethyl 2-amino-4-fluoro-5-methoxybenzoate

  • MDL number MFCD16159160
  • Cas Number : 1247376-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111506
Availability details for
Quantity
  • Availability
  • Add to Cart