ENA372572828 Enamine

N-(6-aminopyridin-3-yl)-4-methoxybenzamide

  • MDL number MFCD18038199
  • Cas Number : 1290702-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111507
Availability details for
Quantity
  • Availability
  • Add to Cart