ENA372572866 Enamine

4-amino-1-cyclohexylpyrrolidin-2-one hydrochloride

  • MDL number MFCD11560322
  • Cas Number : 1177347-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111599
Availability details for
Quantity
  • Availability
  • Add to Cart