ENA372572868 Enamine

4-(aminomethyl)-2-bromophenol hydrobromide

  • MDL number MFCD22392117
  • Cas Number : 63491-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111601
Availability details for
Quantity
  • Availability
  • Add to Cart