ENA372572872 Enamine

(cyanomethyl)urea

  • MDL number MFCD03093030

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111605
Availability details for
Quantity
  • Availability
  • Add to Cart