ENA372572873 Enamine

5-(2,2,2-trifluoroethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD12104394
  • Cas Number : 1154677-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111606
Availability details for
Quantity
  • Availability
  • Add to Cart