ENA372572874 Enamine

methyl 4,5-dihydro-1H-imidazole-4-carboxylate hydrochloride

  • MDL number MFCD22392119
  • Cas Number : 1394729-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111613
Availability details for
Quantity
  • Availability
  • Add to Cart