ENA372572880 Enamine

1-cyanoethyl benzoate

  • MDL number MFCD22392120
  • Cas Number : 3478-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111620
Availability details for
Quantity
  • Availability
  • Add to Cart