ENA372572884 Enamine

4-(aminomethyl)-N,N-dimethylpiperidine-1-sulfonamide hydrochloride

  • MDL number MFCD22392121
  • Cas Number : 1394702-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111627
Availability details for
Quantity
  • Availability
  • Add to Cart