ENA372572885 Enamine

3-chloro-6-(4,5-dimethyl-1H-imidazol-1-yl)pyridazine

  • MDL number MFCD16715816
  • Cas Number : 1248926-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111628
Availability details for
Quantity
  • Availability
  • Add to Cart