ENA372572887 Enamine

2-(chloromethyl)-6-(propan-2-yloxy)pyridine

  • MDL number MFCD16697949
  • Cas Number : 1249448-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111630
Availability details for
Quantity
  • Availability
  • Add to Cart