ENA372572894 Enamine

2-methyl-N1-[(1H-1,2,3,4-tetrazol-5-yl)methyl]benzene-1,3-diamine

  • MDL number MFCD17285139
  • Cas Number : 1341643-44-4
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-N~1~-(1H-tetraazol-5-ylmethyl)-1,3-benzenediamine
Inchi Code 1S/C9H12N6/c1-6-7(10)3-2-4-8(6)11-5-9-12-14-15-13-9/h2-4,11H,5,10H2,1H3,(H,12,13,14,15)
Inchi Key JAUPUYPLBZMVEX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-111655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111655
Availability details for
Quantity
  • Availability
  • Add to Cart