ENA372572895 Enamine

3-(pyrrolidine-1-sulfonyl)pyrrolidine hydrochloride

  • MDL number MFCD22392124
  • Cas Number : 1394704-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111656
Availability details for
Quantity
  • Availability
  • Add to Cart