ENA372572901 Enamine

3-fluoro-4-(morpholin-4-yl)benzonitrile

  • MDL number MFCD03168028
  • Cas Number : 588708-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107998
Availability details for
Quantity
  • Availability
  • Add to Cart