ENA372572902 Enamine

2-(chloromethyl)-1-benzofuran

  • MDL number MFCD00052740
  • Cas Number : 36754-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111666
Availability details for
Quantity
  • Availability
  • Add to Cart