ENA372572909 Enamine

5-fluoro-3-methyl-1-benzofuran-2-sulfonyl chloride

  • MDL number MFCD22033673
  • Cas Number : 1369140-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108010
Availability details for
Quantity
  • Availability
  • Add to Cart