ENA372572919 Enamine

2-(4-aminopiperidin-1-yl)cyclohexan-1-ol

  • MDL number MFCD12801859
  • Cas Number : 1184360-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111716
Availability details for
Quantity
  • Availability
  • Add to Cart