ENA372572922 Enamine

4-bromo-5-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD14657923
  • Cas Number : 60061-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111730

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111730
Availability details for
Quantity
  • Availability
  • Add to Cart