ENA372572928 Enamine

2-(2-hydroxyphenyl)acetamide

  • MDL number MFCD00025519
  • Cas Number : 22446-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-111736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111736
Availability details for
Quantity
  • Availability
  • Add to Cart