ENA372572945 Enamine

5-(methoxymethyl)-3-methyl-1H-1,2,4-triazole

  • MDL number MFCD14611319
  • Cas Number : 89417-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111847
Availability details for
Quantity
  • Availability
  • Add to Cart