ENA372572950 Enamine

6-cyclopropyl-2-methylpyrimidin-4-amine

  • MDL number MFCD12032929
  • Cas Number : 1159818-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111856
Availability details for
Quantity
  • Availability
  • Add to Cart