ENA372572953 Enamine

methyl 4-(3-methyl-1,2,4-oxadiazol-5-yl)butanoate

  • MDL number MFCD22392138
  • Cas Number : 1394683-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111861

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111861
Availability details for
Quantity
  • Availability
  • Add to Cart