ENA372572957 Enamine

5-(4-chloro-3-ethylphenoxy)pyrazine-2-carbonitrile

  • MDL number MFCD20153090
  • Cas Number : 1394791-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111870
Availability details for
Quantity
  • Availability
  • Add to Cart