ENA372572960 Enamine

2-(4-methoxybenzoyl)pyrrolidine hydrochloride

  • MDL number MFCD22392139
  • Cas Number : 1394745-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111873
Availability details for
Quantity
  • Availability
  • Add to Cart