ENA372572961 Enamine

1-[(1-methylpyrrolidin-3-yl)methyl]-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD12165652
  • Cas Number : 1182947-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-111874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111874
Availability details for
Quantity
  • Availability
  • Add to Cart