ENA372572966 Enamine

1-[3-(1H-imidazol-1-yl)propyl]imidazolidine-2,4,5-trione hydrochloride

  • MDL number MFCD22392142
  • Cas Number : 1394703-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111880

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111880
Availability details for
Quantity
  • Availability
  • Add to Cart