ENA372572968 Enamine

ethyl 1,5-dimethyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD11557239
  • Cas Number : 37670-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108100
Availability details for
Quantity
  • Availability
  • Add to Cart