ENA372572972 Enamine

2-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD18729909
  • Cas Number : 941594-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111932

Safety Information

signalword Warning
pcodes P304+P312,P312
MSDS https://www.enaminestore.com/catalog/EN300-111932
hstatements H303,H313,H333
Availability details for
Quantity
  • Availability
  • Add to Cart