ENA372572973 Enamine

2-(benzylamino)-1-(3-methylphenyl)ethan-1-ol

  • MDL number MFCD11919964
  • Cas Number : 94673-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111935

Safety Information

msds https://www.enaminestore.com/catalog/EN300-111935
Availability details for
Quantity
  • Availability
  • Add to Cart