ENA372572986 Enamine

6-bromo-8-chloro-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD20395469
  • Cas Number : 1342961-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111953
Availability details for
Quantity
  • Availability
  • Add to Cart