ENA372572998 Enamine

octahydroindolizin-7-one

  • MDL number MFCD17013152
  • Cas Number : 2407-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108455
Availability details for
Quantity
  • Availability
  • Add to Cart