ENA372573000 Enamine

9-[(thiophen-2-yl)methyl]-9-azabicyclo[3.3.1]nonan-3-one

  • MDL number MFCD16854932
  • Cas Number : 1341744-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111981
Availability details for
Quantity
  • Availability
  • Add to Cart