ENA372573003 Enamine

6-(morpholin-4-yl)pyridine-3-carboximidamide dihydrochloride

  • MDL number MFCD22392152
  • Cas Number : 1394670-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-111991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-111991
Availability details for
Quantity
  • Availability
  • Add to Cart