ENA372573014 Enamine

6-cyclopropylimidazo[2,1-b][1,3]thiazole-5-carbaldehyde

  • MDL number MFCD05179685
  • Cas Number : 564443-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112016

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112016
Availability details for
Quantity
  • Availability
  • Add to Cart