ENA372573018 Enamine

2-ethyl-1,2,3,4-tetrahydroisoquinolin-5-amine

  • MDL number MFCD12821598
  • Cas Number : 223700-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112020
Availability details for
Quantity
  • Availability
  • Add to Cart