ENA372573021 Enamine

1-{[1,2,3,4]tetrazolo[1,5-a]pyrazin-5-yl}piperazine

  • MDL number MFCD16112961
  • Cas Number : 1250756-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112026
Availability details for
Quantity
  • Availability
  • Add to Cart