ENA372573024 Enamine

(7-chloro-2H-1,3-benzodioxol-5-yl)methanamine

  • MDL number MFCD09054834
  • Cas Number : 760936-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112029
Availability details for
Quantity
  • Availability
  • Add to Cart