ENA372573025 Enamine

3-(2-iodoacetyl)-1,3-oxazolidin-2-one

  • MDL number MFCD20415293
  • Cas Number : 444799-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112030
Availability details for
Quantity
  • Availability
  • Add to Cart