ENA372573028 Enamine

4-bromo-2-methanesulfonyl-1-methoxybenzene

  • MDL number MFCD16250388
  • Cas Number : 90531-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112033
Availability details for
Quantity
  • Availability
  • Add to Cart