ENA372573029 Enamine

2-chloro-4-acetamidobenzoic acid

  • MDL number MFCD03756645
  • Cas Number : 38667-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-112035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112035
Availability details for
Quantity
  • Availability
  • Add to Cart