ENA372573031 Enamine

2-(6-bromo-1H-indol-3-yl)acetonitrile

  • MDL number MFCD09954794
  • Cas Number : 152213-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112038
Availability details for
Quantity
  • Availability
  • Add to Cart