ENA372573033 Enamine

N-[4-(carbamimidoylmethoxy)phenyl]-2-methylpropanamide hydrochloride

  • MDL number MFCD22392156
  • Cas Number : 1394668-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112041

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112041
Availability details for
Quantity
  • Availability
  • Add to Cart