ENA372573038 Enamine

4-methyl-3-phenyl-4H-1,2,4-triazole

  • MDL number MFCD11844933
  • Cas Number : 3357-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112050

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112050
Availability details for
Quantity
  • Availability
  • Add to Cart