ENA372573039 Enamine

1-(3-bromopropyl)piperidine hydrobromide

  • MDL number MFCD13176495
  • Cas Number : 58689-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-205
Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112074
Availability details for
Quantity
  • Availability
  • Add to Cart