ENA372573042 Enamine

N'-hydroxy-2-methoxy-4,6-dimethylpyridine-3-carboximidamide

  • Cas Number : 1251436-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112077
Availability details for
Quantity
  • Availability
  • Add to Cart